Skip to Content
Merck
All Photos(1)

Key Documents

290149

Sigma-Aldrich

2,4-Bis(trifluoromethyl)benzyl bromide

95%

Synonym(s):

1-(Bromomethyl)-2,4-bis(trifluoromethyl)benzene

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$83.60
5 G
$354.00

$83.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$83.60
5 G
$354.00

About This Item

Linear Formula:
(CF3)2C6H3CH2Br
CAS Number:
Molecular Weight:
307.03
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$83.60


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

refractive index

n20/D 1.451 (lit.)

density

1.637 g/mL at 25 °C (lit.)

functional group

bromo
fluoro

SMILES string

FC(F)(F)c1ccc(CBr)c(c1)C(F)(F)F

InChI

1S/C9H5BrF6/c10-4-5-1-2-6(8(11,12)13)3-7(5)9(14,15)16/h1-3H,4H2

InChI key

SWFFFUJOWAJJCH-UHFFFAOYSA-N

Application

2,4-Bis(trifluoromethyl)benzyl bromide may be used in chemical synthesis studies.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

129.2 °F - closed cup

Flash Point(C)

54 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service