Skip to Content
Merck
All Photos(1)

Documents

287768

Sigma-Aldrich

Silicon phthalocyanine dichloride

Dye content ~85 %

Synonym(s):

Dichloro(29H,31H-phthalocyaninato)silicon, Phthalocyaninato-dichlorosilane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H16Cl2N8Si
CAS Number:
Molecular Weight:
611.51
Beilstein:
600612
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

powder

composition

Dye content, ~85%

λmax

702 nm

SMILES string

Cl[Si]1(Cl)n2c3nc4nc(nc5n1c(nc6nc(nc2c7ccccc37)c8ccccc68)c9ccccc59)c%10ccccc4%10

InChI

1S/C32H16Cl2N8Si/c33-43(34)41-29-21-13-5-6-14-22(21)31(41)39-27-19-11-3-4-12-20(19)28(36-27)40-32-24-16-8-7-15-23(24)30(42(32)43)38-26-18-10-2-1-9-17(18)25(35-26)37-29/h1-16H/b37-25-,37-29-,38-26-,38-30-,39-27-,39-31-,40-28-,40-32-

InChI key

NURIJECXIAPSAM-MLODSWNHSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Precursor to stacked-ring silicon phthalocyanines.

Other Notes

remainder SiO2

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of the American Chemical Society, 107, 192-192 (1985)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service