Skip to Content
Merck
All Photos(2)

Key Documents

379557

Sigma-Aldrich

Manganese(II) phthalocyanine

Synonym(s):

Phthalocyanine manganese(II) salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$79.50

$79.50


Estimated to ship on27 March 2025


Request a Bulk Order

Select a Size

Change View
1 G
$79.50

About This Item

Empirical Formula (Hill Notation):
C32H16MnN8
CAS Number:
Molecular Weight:
567.46
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

$79.50


Estimated to ship on27 March 2025


Request a Bulk Order

form

powder

Quality Level

λmax

727 nm

SMILES string

[Mn]1n2c3nc4nc(nc5n1c(nc6nc(nc2c7ccccc37)c8ccccc68)c9ccccc59)c%10ccccc4%10

InChI

1S/C32H16N8.Mn/c1-2-10-18-17(9-1)25-33-26(18)38-28-21-13-5-6-14-22(21)30(35-28)40-32-24-16-8-7-15-23(24)31(36-32)39-29-20-12-4-3-11-19(20)27(34-29)37-25;/h1-16H;/q-2;+2

InChI key

ICIFYHOILPYQKB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Manganese (II) phthalocyanine is used as a coating of quartz crystals which is to be used as sensors for detecting H2S.[1] It is used to discriminate between cheese varieties,[2] detect volatile hazardous compounds[3] and spoilt fruit.[4]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new analytical system, based on an acoustic wave sensor, for halitosis evaluation.
Fernandes DLA, et al.
Sensors and Actuators B, Chemical, 136(1), 73-79 (2009)
Detecting spoiled fruit in the house of the future
Fernandes DLA, et al.
Analytica Chimica Acta, 617(1), 171-176 (2008)
Daniel L A Fernandes et al.
Talanta, 77(1), 77-83 (2008-09-23)
A new electronic nose was developed to identify the chemical compound released when a 2.5-L flask was broken inside a 3 m x 3 m x 2.5 m store-room. Flasks of 10 different hazardous compounds were initially present in the
Jan Kruid et al.
Chemosphere, 175, 247-252 (2017-02-23)
Identifying the most efficient oxidation process to achieve maximum removal of a target pollutant compound forms the subject of much research. There exists a need to develop rapid screening tools to support research in this area. In this work we
Liang Huang et al.
Science advances, 5(5), eaav5490-eaav5490 (2019-05-07)
Conventional nanozyme technologies face formidable challenges of intricate size-, composition-, and facet-dependent catalysis and inherently low active site density. We discovered a new class of single-atom nanozymes with atomically dispersed enzyme-like active sites in nanomaterials, which significantly enhanced catalytic performance

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service