Skip to Content
Merck
All Photos(1)

Key Documents

285366

Sigma-Aldrich

Ethyl 2-(4-chlorophenoxy)-2-methylpropionate

99%

Synonym(s):

2-(p-Chlorophenoxy)-2-methylpropionic acid ethyl ester, Clofibrate, Ethyl 2-(p-chlorophenoxy)isobutyrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15ClO3
CAS Number:
Molecular Weight:
242.70
Beilstein:
1913459
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

Assay

99%

bp

154-156 °C/20 mmHg (lit.)

density

1.14 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)C(C)(C)Oc1ccc(Cl)cc1

InChI

1S/C12H15ClO3/c1-4-15-11(14)12(2,3)16-10-7-5-9(13)6-8-10/h5-8H,4H2,1-3H3

InChI key

KNHUKKLJHYUCFP-UHFFFAOYSA-N

Gene Information

human ... PPARA(5465)

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masayuki Yoshikawa et al.
Journal of natural products, 68(9), 1360-1365 (2005-09-27)
The methanolic extract and its n-butanol-soluble fraction from the flowers of the tea plant (Camellia sinensis) were found to suppress serum triglyceride elevation in olive oil-treated mice. From the n-butanol-soluble fraction, three new acylated oleanane-type triterpene oligoglycosides, floratheasaponins A-C (1-3)
Sara Chuang et al.
Journal of medicinal chemistry, 52(17), 5344-5355 (2009-08-12)
Liver-fatty acid binding protein (L-FABP) is found in high levels in enterocytes and is involved in cytosolic solubilization of fatty acids. In addition, L-FABP has been shown to bind endogenous and exogenous lipophilic compounds, suggesting that it may also play
Toshio Morikawa et al.
Journal of natural products, 71(5), 828-835 (2008-03-28)
The methanolic extract and its saponin fraction (methanol-eluted fraction) of the flowers of Bellis perennis were found to suppress serum triglyceride elevation in olive oil-treated mice. From the saponin fraction, seven new triterpene saponins, perennisosides I (1), II (2), III
H M Mehendale
Toxicological sciences : an official journal of the Society of Toxicology, 57(2), 187-190 (2000-09-28)
The article highlighted in this issue is "Peroxisome Proliferator-Activated Receptor Alpha-Null Mice Lack Resistance to Acetaminophen Hepatotoxicity Following Clofibrate Exposure" by Chuan Chen, Gayle E. Hennig, Herbert E. Whiteley, J Christopher Corton, and José E. Manautou (pp. 338-344).
Clofibrate myopathy: a case report and a review of the literature.
P Rush et al.
Seminars in arthritis and rheumatism, 15(3), 226-229 (1986-02-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service