Skip to Content
Merck
All Photos(1)

Key Documents

284483

Sigma-Aldrich

(R)-(−)-2-Amino-3-methyl-1-butanol

98%

Synonym(s):

D-Valinol

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$48.50
5 G
$141.00

$48.50


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$48.50
5 G
$141.00

About This Item

Linear Formula:
(CH3)2CHCH(NH2)CH2OH
CAS Number:
Molecular Weight:
103.16
Beilstein:
1719138
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

$48.50


Check Cart for Availability

Request a Bulk Order

Assay

98%

form

solid

optical activity

[α]20/D −16°, c = 10 in ethanol

mp

35-36 °C (lit.)

functional group

amine
hydroxyl

SMILES string

CC(C)[C@@H](N)CO

InChI

1S/C5H13NO/c1-4(2)5(6)3-7/h4-5,7H,3,6H2,1-2H3/t5-/m0/s1

InChI key

NWYYWIJOWOLJNR-YFKPBYRVSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

172.4 °F - closed cup

Flash Point(C)

78 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service