Skip to Content
Merck
All Photos(2)

Key Documents

282626

Sigma-Aldrich

Trimethoxysilane

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)3SiH
CAS Number:
Molecular Weight:
122.20
Beilstein:
1697990
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

vapor density

>1 (vs air)

vapor pressure

<7.2 mmHg ( 20 °C)

Assay

95%

form

liquid

refractive index

n20/D 1.358 (lit.)

bp

81 °C (lit.)

mp

−115 °C (lit.)

density

0.96 g/mL at 25 °C (lit.)

SMILES string

CO[SiH](OC)OC

InChI

1S/C3H10O3Si/c1-4-7(5-2)6-3/h7H,1-3H3

InChI key

YUYCVXFAYWRXLS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

48.2 °F - closed cup

Flash Point(C)

9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chen-Cheng Xue et al.
Science advances, 6(18), eaax1346-eaax1346 (2020-06-05)
The rapid development of treatment resistance in tumors poses a technological bottleneck in clinical oncology. Ferroptosis is a form of regulated cell death with clinical translational potential, but the efficacy of ferroptosis-inducing agents is susceptible to many endogenous factors when
H Cheraghi Bidsorkhi et al.
Nanotechnology, 27(41), 415706-415706 (2016-09-09)
In this research, novel ultrafiltration nanocomposite membranes were prepared by incorporating self-synthesized nanoporous titanium dioxide (NTiO2) nanoparticles into polysulfone. The surface of the nanoparticle was treated with a silane-based modifier to improve its distribution in the host polymer. Atomic-force microscopy
Wonryung Lee et al.
Advanced materials (Deerfield Beach, Fla.), 28(44), 9722-9728 (2016-10-08)
Integration of organic electrochemical transistors and organic field-effect transistors is successfully realized on a 600 nm thick parylene film toward an electrophysiology array. A single cell of an integrated device and a 2 × 2 electrophysiology array succeed in detecting
Othman Bouloussa et al.
Chemical communications (Cambridge, England), (8)(8), 951-953 (2008-02-20)
A new, simple method to obtain ultrathin polycationic monolayers on hydroxylated surfaces is described which uses a bifunctional copolymer comprising a reactive part (trimethoxysilane) and positive charges (quaternary ammonium salts) to confer antimicrobial properties.
Mingquan Liu et al.
Analytical chemistry, 91(15), 10188-10196 (2019-07-10)
Reversible protein phosphorylation on serine, threonine, and tyrosine residues is essential for fast, specific, and accurate signal transduction in cells. Up to now, the identification and quantification of phosphorylated amino acids, peptides, and proteins continue to be one of the

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service