Skip to Content
Merck
All Photos(1)

Documents

66612

Sigma-Aldrich

Methyldiethoxysilane

≥96%

Synonym(s):

Diethoxy-methylsilane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H14O2Si
CAS Number:
Molecular Weight:
134.25
Beilstein:
1733975
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥96%

bp

94-95 °C (lit.)

density

0.838 g/mL at 20 °C (lit.)

SMILES string

CCO[SiH](C)OCC

InChI

1S/C5H14O2Si/c1-4-6-8(3)7-5-2/h8H,4-5H2,1-3H3

InChI key

SOGIFFQYRAXTDR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

33.8 °F - closed cup

Flash Point(C)

1 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Bhushan P Mahadik et al.
Biomaterials, 67, 297-307 (2015-08-04)
Hematopoietic stem cells (HSCs) are a rare stem cell population found primarily in the bone marrow and responsible for the production of the body's full complement of blood and immune cells. Used clinically to treat a range of hematopoietic disorders

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service