Skip to Content
Merck
All Photos(1)

Key Documents

281905

Sigma-Aldrich

o-Tolylhydrazine hydrochloride

97%

Synonym(s):

2-Methylphenylhydrazine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4NHNH2 · HCl
CAS Number:
Molecular Weight:
158.63
Beilstein:
3655929
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
form:
powder
Assay:
97%

Assay

97%

form

powder

mp

190 °C (dec.) (lit.)

solubility

alcohol: soluble
chloroform: soluble
diethyl ether: soluble
petroleum ether: slightly soluble (cold)
water: very slightly soluble

functional group

hydrazine

SMILES string

Cl.Cc1ccccc1NN

InChI

1S/C7H10N2.ClH/c1-6-4-2-3-5-7(6)9-8;/h2-5,9H,8H2,1H3;1H

InChI key

KJGFNDCSTWGUDT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

o-Tolylhydrazine hydrochloride has been used in the preparation of o-tolylhydrazone of D-galactose.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The detection of galactose in urine.
F S FOWWEATHER
The Biochemical journal, 55(4), 718-720 (1953-11-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service