Skip to Content
Merck
All Photos(2)

Key Documents

278602

Sigma-Aldrich

1-Dodecylpyridinium chloride hydrate

98%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$91.30

$91.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$91.30

About This Item

Empirical Formula (Hill Notation):
C17H30ClN · xH2O
CAS Number:
Molecular Weight:
283.88 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

$91.30


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

liquid crystal

mp

66-70 °C (lit.)

SMILES string

[Cl-].[H]O[H].CCCCCCCCCCCC[n+]1ccccc1

InChI

1S/C17H30N.ClH.H2O/c1-2-3-4-5-6-7-8-9-10-12-15-18-16-13-11-14-17-18;;/h11,13-14,16-17H,2-10,12,15H2,1H3;1H;1H2/q+1;;/p-1

InChI key

BDGGUWSWAKGEGH-UHFFFAOYSA-M

General description

Dodecylpyridinium chloride is a cationic surfactant. [1]

Application

Micellization characteristics of dodecylpyridinium chloride have been studied.[2] Properties such as molar conductivity, surface tension and surface adsorption parameters has been reported.[3] A non-ionic-cationic mixed system composed of Triton X-100 and dodecylpyridinium chloride respectively was studied for its clouding behavior.[1]

Legal Information

Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and properties of cationic surfactants with tuned hydrophylicity
Quagliotto P, et al.
Journal of Colloid and Interface Science, 340(2), 269-276 (2009)
Co- and Counterion Effect on the Micellization Characteristics of Dodecylpyridinium Chloride,
Bhat MA, et al.
Journal of Dispersion Science and Technology, 29(4) null
Clouding behavior of nonionic?cationic and nonionic?anionic mixed surfactant systems in presence of carboxylic acids and their sodium salts
Jan M, et al.
Coll. Polymer Sci., 285(6), 631- 640 null
D B França et al.
Chemosphere, 242, 125109-125109 (2019-11-02)
Organoclays have been applied as efficient adsorbents for pharmaceutical pollutants from aqueous solution. In this work, dodecylpyridinium chloride (C12pyCl) and hexadecylpyridinium chloride (C16pyCl) cationic surfactants were used for the preparation of organobentonites destined for diclofenac sodium (DFNa) adsorption, an anionic
Daniel Ondo et al.
Journal of colloid and interface science, 505, 445-453 (2017-06-20)
The interaction of α-cyclodextrin (α-CD) with ten ionic surfactants (S) in water was systematically examined using isothermal titration calorimetry. The S comprised cationic and anionic head groups while the hydrocarbon alkyl chain length varied from eight to fourteen carbon atoms.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service