All Photos(1)
About This Item
Linear Formula:
F2C6H3COCH3
CAS Number:
Molecular Weight:
156.13
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
refractive index
n20/D 1.4882 (lit.)
density
1.233 g/mL at 25 °C (lit.)
functional group
fluoro
ketone
SMILES string
CC(=O)c1cc(F)ccc1F
InChI
1S/C8H6F2O/c1-5(11)7-4-6(9)2-3-8(7)10/h2-4H,1H3
InChI key
HLAFIZUVVWJAKL-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
2′,5′-Difluoroacetophenone has been used in the preparation of 2,5-difluorostyrene.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
2, 3-Difluorostyrene and 2-Chloro-5-fluorostyrene. The Preparation of Aromatic Fluorine Compounds.
BERGMANN ED and Berkovic S.
The Journal of Organic Chemistry, 26(3), 919-923 (1961)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service