Skip to Content
Merck
All Photos(1)

Documents

261211

Sigma-Aldrich

Samarium

chips, 99.9% trace rare earth metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Sm
CAS Number:
Molecular Weight:
150.36
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9% trace rare earth metals basis

form

chips

reaction suitability

reagent type: catalyst
core: samarium

resistivity

91.4 μΩ-cm, 0°C

bp

1794 °C (lit.)

mp

1074 °C (lit.)

density

7.47 g/mL at 25 °C (lit.)

SMILES string

[Sm]

InChI

1S/Sm

InChI key

KZUNJOHGWZRPMI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi Lu et al.
Optics express, 21(2), 1997-2002 (2013-02-08)
We demonstrate an all-fiber passively Q-switched Yb-doped laser using a piece of Sm-doped fiber as a saturable absorber. The laser was pumped by two 25W, 975 nm fiber coupled diodes and Q-switching was initiated when the ASE generated in the
M Majumder et al.
Journal of physics. Condensed matter : an Institute of Physics journal, 25(2), 025701-025701 (2012-12-05)
We report resistivity, magnetization and (19)F NMR results in a polycrystalline sample of SmFeAsO(0.86)F(0.14). The resistivity and magnetization data show a sharp drop at 48 K indicating a superconducting transition. The nuclear spin-lattice rate (1/T(1)) and spin-spin relaxation rate (1/T(2))
I Pal et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 101, 74-81 (2012-10-27)
The glasses with compositions 20ZnO·(79.5-x)Bi2O3·xSiO2·0.5Sm2O3 (10≤x≤50, mol%) have been synthesized using normal melt-quench technique. Optical absorption and fluorescence spectra of the glasses were recorded at ambient temperature. Judd-Ofelt (J-O) theory has been successfully applied to characterize the absorption and luminescence
Hua Yang et al.
Inorganic chemistry, 52(3), 1275-1284 (2013-01-25)
Sm@C(2v)(3)-C(80) has been separated from the carbon soot produced by electrical arc vaporization of graphite rods doped with Sm(2)O(3) and purified. Its structure has been determined by single crystal X-ray diffraction using cocrystals obtained from either Ni(II)(octaethylporphyrin) (Ni(II)(OEP)) to form
Benjamin Voß et al.
Nanoscale, 5(2), 806-812 (2012-12-14)
The origin of the narrow particle size distributions obtained in the oleic acid-based synthesis of hexagonal phase β-NaREF(4) nanocrystals (RE = Sm, Eu, Gd, Tb) has been investigated. Compared to the standard synthesis, the growth conditions were simplified by using

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service