Skip to Content
Merck
All Photos(1)

Key Documents

260614

Sigma-Aldrich

Ethyl 3-benzoylacrylate

technical grade, 92%

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
$163.00

$163.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 ML
$163.00

About This Item

Linear Formula:
C6H5COCH=CHCO2C2H5
CAS Number:
Molecular Weight:
204.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$163.00


Check Cart for Availability

Request a Bulk Order

grade

technical grade

Quality Level

Assay

92%

form

liquid

refractive index

n20/D 1.543 (lit.)

bp

184-185 °C/25 mmHg (lit.)

density

1.112 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)\C=C\C(=O)c1ccccc1

InChI

1S/C12H12O3/c1-2-15-12(14)9-8-11(13)10-6-4-3-5-7-10/h3-9H,2H2,1H3/b9-8+

InChI key

ACXLBHHUHSJENU-CMDGGOBGSA-N

General description

Ethyl 3-benzoylacrylate (ethyl trans-3-benzoylacrylate) undergoes enantioselective guanidine catalyst promoted Michael addition reaction with dithranol to yield Michael adduct[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juan Shen et al.
Organic & biomolecular chemistry, 6(18), 3229-3236 (2008-09-20)
The enantioselective Diels-Alder reaction is one of the most important reactions for the synthesis of complex molecules. It provides access to chiral six-membered carbocyclic compounds containing up to four stereogenic centers in a single step. Asymmetric catalysis in the Diels-Alder

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service