Skip to Content
Merck
All Photos(1)

Key Documents

304190

Sigma-Aldrich

2-Methyl-2-cyclopenten-1-one

98%

Synonym(s):

2-Methyl-2-cyclopentene-1-one, 2-Methyl-2-cyclopentenone

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$130.00
5 G
$328.00

$130.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$130.00
5 G
$328.00

About This Item

Linear Formula:
CH3C5H5(=O)
CAS Number:
Molecular Weight:
96.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$130.00


Check Cart for Availability

Request a Bulk Order

Assay

98%

form

liquid

refractive index

n20/D 1.479 (lit.)

bp

158-161 °C (lit.)

density

0.979 g/mL at 25 °C (lit.)

functional group

ketone

storage temp.

2-8°C

SMILES string

CC1=CCCC1=O

InChI

1S/C6H8O/c1-5-3-2-4-6(5)7/h3H,2,4H2,1H3

InChI key

ZSBWUNDRDHVNJL-UHFFFAOYSA-N

General description

Diels-Alder reactions of 2-methyl-2-cyclopenten-1-one with 1-ethenylhydronaphthalenes were studied.[1]

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

120.2 °F - closed cup

Flash Point(C)

49 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regiochemistry of the Diels-Alder Reaction of 1-Ethenyl-hydronaphthalenes and 2-Methyl-2-cyclopenten-1-one.
Minuti L, et al.
Synthetic Communications, 22(11), 1535-1540 (1992)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service