252794
Methyl 3-hydroxybenzoate
99%
Synonym(s):
3-(Methoxycarbonyl)phenol, 3-Carbomethoxyphenol, 3-Hydroxybenzoic acid methyl ester, Methyl m-hydroxybenzoate, m-Carbomethoxyphenol, m-Hydroxybenzoic acid methyl ester
About This Item
Recommended Products
Assay
99%
form
solid
bp
280-281 °C/709 mmHg (lit.)
mp
70-72 °C (lit.)
SMILES string
COC(=O)c1cccc(O)c1
InChI
1S/C8H8O3/c1-11-8(10)6-3-2-4-7(9)5-6/h2-5,9H,1H3
InChI key
YKUCHDXIBAQWSF-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
- 3-hydroxybenzene-1,2-dicarbaldehyde
- O-methyl O-[3-methyl-4-(methylthio)phenyl]O-(3-methylcarboxyphenyl) phosphorothioate
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service