Skip to Content
Merck
All Photos(2)

Key Documents

250929

Sigma-Aldrich

N,N′-Bis(salicylidene)-1,3-propanediamine

99%

Synonym(s):

N,N′-Disalicylidene-1,3-diaminopropane, N,N′-Trimethylene-bis(salicylimine)

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$160.00

$160.00


Estimated to ship on15 April 2025


Request a Bulk Order

Select a Size

Change View
25 G
$160.00

About This Item

Linear Formula:
CH2(CH2N=CHC6H4OH)2
CAS Number:
Molecular Weight:
282.34
Beilstein:
2057483
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$160.00


Estimated to ship on15 April 2025


Request a Bulk Order

Assay

99%

form

solid

mp

51-53 °C (lit.)

functional group

amine
imine

SMILES string

Oc1ccccc1\C=N\CCC\N=C\c2ccccc2O

InChI

1S/C17H18N2O2/c20-16-8-3-1-6-14(16)12-18-10-5-11-19-13-15-7-2-4-9-17(15)21/h1-4,6-9,12-13,20-21H,5,10-11H2/b18-12+,19-13+

InChI key

KLDZYURQCUYZBL-KLCVKJMQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,N′-Bis(salicylidene)-1,3-propanediamine is a di-Schiff base ligand[1].

Application

N,N′-Bis(salicylidene)-1,3-propanediamine has been used in the synthesis of:
  • new trinuclear heterometallic Cu(II)-Mn(II) complexes[1]
  • mononuclear nickel complex, NiL (LH2 = N,N′-bis(salicylidene)-1,3-diaminopropane)[2]
Can be used as a selective chelating titrant for Cu(II)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas Radoske et al.
Dalton transactions (Cambridge, England : 2003), 49(48), 17559-17570 (2020-11-21)
A series of tetradentate N2O2-type Schiff base complexes with tetravalent 4f- and 5f-block metals, [M(salpn)2] (H2salpn = N,N'-disalicylidene-1,3-diaminopropane; M = Ce, Th, U, Np, and Pu), were prepared to systematically investigate their solid state structure, and their complexation behaviour in
Piya Seth et al.
Dalton transactions (Cambridge, England : 2003), 43(3), 990-998 (2013-10-29)
Five new trinuclear heterometallic Cu(II)-Mn(II) complexes [(CuL)2Mn(O2CPh)2] (1), [(CuL)2Mn(N3)2] (2), [(CuL)2Mn(NCO)2] (3), [(CuL)2Mn(NO3)2] (4) and [(CuL)2Mn(Sal)2]·CH2Cl2 (5) have been synthesized with the di-Schiff base ligand H2L (where H2L = N,N'-bis(salicylidene)-1,3-propanediamine and Sal = salicylate). These complexes with different anionic co-ligands
Raif Kurtaran et al.
Journal of inorganic biochemistry, 99(10), 1937-1944 (2005-08-02)
The mononuclear nickel complex NiL (LH2 = N,N'-bis(salicylidene)-1,3-diaminopropane) can be transformed into the tetranuclear complex [NiL(H2O)Pb(SCN)2(DMF)]2 in the aid of SCN-, DMF (dimethylformamide) and Pb(II) ions. The complex was characterized by elemental analysis and FTIR investigation. The crystal structure reveals
O Bergendorff et al.
Contact dermatitis, 42(1), 11-17 (2000-01-22)
N,N'-disalicylidene-1,2-diaminopropane is a copper inhibitor present in some adhesive plasters, rubber products and gasoline. Upon contact with water it is hydrolyzed to salicylaldehyde and 1,2-diaminopropane. All patients in this study showed positive patch-test reactions to N,N'-disalicylidene-1,2-diaminopropane, and also to 1,2-diaminopropane

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service