Skip to Content
Merck
All Photos(5)

Key Documents

D12384

Sigma-Aldrich

3,3′-Diaminobenzidine

97%

Synonym(s):

3,3′,4,4′-Biphenyltetramine, 3,3′,4,4′-Tetraaminobiphenyl, DAB

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH2)2C6H3C6H3(NH2)2
CAS Number:
Molecular Weight:
214.27
Beilstein:
1212988
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
powder
Assay:
97%

Quality Level

Assay

97%

form

powder

mp

175-177 °C (lit.)

SMILES string

NC1=C(N)C=CC(C2=CC(N)=C(N)C=C2)=C1

InChI

1S/C12H14N4/c13-9-3-1-7(5-11(9)15)8-2-4-10(14)12(16)6-8/h1-6H,13-16H2

InChI key

HSTOKWSFWGCZMH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3,3′-Diaminobenzidine can be used as a starting material to prepare:
  • Aromatic poly(imide) networks for hydrogen storage applications.
  • Various derivatives of quinoxalines by condensation with diketones.
  • Mesitylene-containing poly(benzimidazolium) analogs (Mes-PBI) for alkali anion-exchange membrane.
Peroxidase substrate; reagent for spectrophotometric determination of selenium.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Muta. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A stable hydroxide-conducting polymer.
Thomas OD, et al.
Journal of the American Chemical Society, 134(26), 10753-10756 (2012)
n-Type conjugated oligoquinoline and oligoquinoxaline with triphenylamine endgroups: efficient ambipolar light emitters for device applications.
Hancock JM, et al.
Chemistry of Materials, 18(20), 4924-4932 (2006)
Microporous networks of high-performance polymers: Elastic deformations and gas sorption properties.
Weber J, et al.
Macromolecules, 41(8), 2880-2885 (2008)
Muqing Cao et al.
Pediatric research, 78(2), 137-144 (2015-04-29)
Preterm infants show delayed development of motor function after birth. This may relate to functional immaturity of many organs, including the gut and brain. Using pigs as model for preterm infants, we hypothesized that early initiation of enteral feeding stimulates
Fatal human rabies due to Duvenhage virus from a bat in Kenya: failure of treatment with coma-induction, ketamine, and antiviral drugs.
Pieter-Paul A M van Thiel et al.
PLoS neglected tropical diseases, 3(7), e428-e428 (2009-07-29)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service