Skip to Content
Merck
All Photos(1)

Key Documents

250856

Sigma-Aldrich

(1S,2S,3S,5R)-(+)-Isopinocampheol

98%

Synonym(s):

(1S,2S,3S,5R)-2,6,6-Trimethylbicyclo[3.1.1]heptan-3-ol, (1S,2S,3S,5R)-3-Pinanol, 3-Pinanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
Beilstein:
2038104
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

Assay

98%

form

solid

optical activity

[α]20/D +35.1°, c = 20 in ethanol

bp

219 °C (lit.)

mp

51-53 °C (lit.)

SMILES string

C[C@@H]1[C@@H](O)CC2CC1C2(C)C

InChI

1S/C10H18O/c1-6-8-4-7(5-9(6)11)10(8,2)3/h6-9,11H,4-5H2,1-3H3/t6-,7+,8-,9-/m0/s1

InChI key

REPVLJRCJUVQFA-KZVJFYERSA-N

General description

(1S,2S,3S,5R)-(+)-Isopinocampheol is a chiral terpenol.[1]

Application

(1S,2S,3S,5R)-(+)-Isopinocampheol may be used in the preparation of
  • thermotropic chiral nematic side-chain copolymers[2]
  • ortho-(diphenylphosphino)phenylphosphonous acid di[(1S,2S,3S,5R)-2,6,6-trimethyl-bicyclo[3.1.1]-hept-3-yl]ester, a phosphino-phosphonite ligand[3]
  • O,O´bis[(1S,2S,3S,5R)2,6,6-trimethylbicyclo[3.1.1]hept-3-yl] dithiophosphoric acid, a bioactive compound with antifungal effect[4]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

200.1 °F - closed cup

Flash Point(C)

93.4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New thermotropic chiral nematic copolymers using (1S, 2S, 3S, 5R)-(+)-and (1R, 2R, 3R, 5S)-([minus])-isopinocampheol as building blocks.
Chen SH and Tsai ML.
Macromolecules, 23(24) (1990)
Synthesis of enantiopure C 1 symmetric diphosphines and phosphino-phosphonites with ortho-phenylene backbones.
Kottsieper KW, et al.
Tetrahedron Asymmetry, 12(8), 1159-1169 (2001)
Chiral phosphorus dithio acids derived from (1S, 2S, 3S, 5R)-(+)-isopinocampheol. Synthesis and fungicidal activity.
Nizamov IS, et al.
Russian Chemical Bulletin, 61(12), 2370-2371 (2012)
Terpene Analogues of Dithiophospate Pesticides.
Cherkasov RA, et al.
Phosph. Sulfur Relat. Elem., 186(4), 1003-1004 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service