Skip to Content
Merck
All Photos(1)

Key Documents

188417

Sigma-Aldrich

(1R)-(−)-Myrtenol

95%

Synonym(s):

(1R)-2-Pinen-10-ol, (1R)-6,6-Dimethylbicyclo[3.1.1]hept-2-ene-2-methanol

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$240.00
100 G
$628.00

$240.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$240.00
100 G
$628.00

About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
Beilstein:
2206583
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

$240.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

optical activity

[α]20/D −51°, neat

refractive index

n20/D 1.496 (lit.)

bp

221-222 °C (lit.)

density

0.954 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

CC1(C)[C@H]2CC=C(CO)[C@@H]1C2

InChI

1S/C10H16O/c1-10(2)8-4-3-7(6-11)9(10)5-8/h3,8-9,11H,4-6H2,1-2H3/t8-,9-/m0/s1

InChI key

RXBQNMWIQKOSCS-IUCAKERBSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

154.4 °F - closed cup

Flash Point(C)

68 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service