Skip to Content
Merck
All Photos(1)

Documents

246220

Sigma-Aldrich

4-Chloroindole

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6ClN
CAS Number:
Molecular Weight:
151.59
Beilstein:
114880
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.628 (lit.)

bp

129-130 °C/4 mmHg (lit.)

solubility

ethanol: 50 mg/mL, clear, colorless

density

1.259 g/mL at 25 °C (lit.)

SMILES string

Clc1cccc2[nH]ccc12

InChI

1S/C8H6ClN/c9-7-2-1-3-8-6(7)4-5-10-8/h1-5,10H

InChI key

SVLZRCRXNHITBY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The nitrosation rate of 4-chloroindole and the stability of its nitrosated products were studied.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

H G Tiedink et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 27(11), 723-730 (1989-11-01)
The nitrosation rates of indole-3-acetonitrile, indole-3-carbinol, indole and 4-chloroindole and the stability of their nitrosated products were investigated. Each of the nitrosated indole compounds was directly mutagenic to Salmonella typhimurium TA100 in the following order of potency: 4-chloroindole much greater
Zhong-Liu Wu et al.
The Journal of biological chemistry, 280(49), 41090-41100 (2005-10-11)
The natural product indole is a substrate for cytochrome P450 2A6. Mutagenesis of P450 2A6 was done to expand its capability in the oxidization of bulky substituted indole compounds, which are not substrates for the wild-type enzyme or the double
H G Tiedink et al.
Cell biology and toxicology, 7(4), 371-386 (1991-10-01)
4-chloro-methoxyindole is a naturally occurring compound in Vicia faba which can easily react with nitrite to form a N-nitroso compound. In this in vitro study, the potential genotoxic effects of nitrosated 4-chloro-6-methoxyindole and its structural analogue 4-chloroindole were evaluated for
Zhi-Gang Zhang et al.
Journal of biotechnology, 139(1), 12-18 (2008-11-06)
Cytochrome P450 (P450) 2A6 is able to catalyze indole hydroxylation to form the blue dye indigo. The wild-type P450 2A6 enzyme was randomly mutated throughout the whole open reading frame and screened using 4-chloroindole hydroxylation, a substituted indole selected from
Chaitany Jayprakash Raorane et al.
Biomolecules, 10(8) (2020-08-23)
Multi-drug resistant Acinetobacter baumannii is well-known for its rapid acclimatization in hospital environments. The ability of the bacterium to endure desiccation and starvation on dry surfaces for up to a month results in outbreaks of health care-associated infections. Previously, indole

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service