Skip to Content
Merck
All Photos(1)

Key Documents

473731

Sigma-Aldrich

7-Chloroindole

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$173.00

$173.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$173.00

About This Item

Empirical Formula (Hill Notation):
C8H6ClN
CAS Number:
Molecular Weight:
151.59
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$173.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

bp

90-95 °C/0.25 mmHg (lit.)

mp

55-58 °C (lit.)

functional group

chloro

SMILES string

Clc1cccc2cc[nH]c12

InChI

1S/C8H6ClN/c9-7-3-1-2-6-4-5-10-8(6)7/h1-5,10H

InChI key

WMYQAKANKREQLM-UHFFFAOYSA-N

General description

7-Chloroindole is an indole derivative. It has been synthesized from 2,3-dihydroindole.[1]

Application

7-Chloroindole may be used in the preparation of 1-methyl-7-chloroindole[2] and glycosylated 7-chloroindole-3-acetamide.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Synthesis of rebeccamycin and 11-dechlororebeccamycin.
Faul MM, et al.
The Journal of Organic Chemistry, 64(7), 2465-2470 (1999)
Xin Teng et al.
Bioorganic & medicinal chemistry letters, 15(22), 5039-5044 (2005-09-13)
Necroptosis is a regulated caspase-independent cell death mechanism that results in morphological features resembling necrosis. It can be induced in a FADD-deficient variant of human Jurkat T cells treated with TNF-alpha. 5-(1H-Indol-3-ylmethyl)-2-thiohydantoins and 5-(1H-indol-3-ylmethyl)hydantoins were found to be potent necroptosis
The chemistry of indoles. XXXIX. A facile synthetic method for 7-substituted indoles.
Somei M, et al.
Chemical & Pharmaceutical Bulletin, 35(8), 3146-3154 (1987)
Jeongchan Lee et al.
Nature chemical biology, 17(1), 104-112 (2020-11-04)
Tyrian purple, mainly composed of 6,6'-dibromoindigo (6BrIG), is an ancient dye extracted from sea snails and was recently demonstrated as a biocompatible semiconductor material. However, its synthesis remains limited due to uncharacterized biosynthetic pathways and the difficulty of regiospecific bromination.
Chaitany Jayprakash Raorane et al.
Biomolecules, 10(8) (2020-08-23)
Multi-drug resistant Acinetobacter baumannii is well-known for its rapid acclimatization in hospital environments. The ability of the bacterium to endure desiccation and starvation on dry surfaces for up to a month results in outbreaks of health care-associated infections. Previously, indole

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service