Skip to Content
Merck
All Photos(3)

Key Documents

245232

Sigma-Aldrich

Methyl 2-(aminosulfonyl)benzoate

98%

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$144.00
500 G
$310.00

$144.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$144.00
500 G
$310.00

About This Item

Linear Formula:
H2NSO2C6H4CO2CH3
CAS Number:
Molecular Weight:
215.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$144.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

solid

mp

126-128 °C (lit.)

functional group

ester

SMILES string

COC(=O)c1ccccc1S(N)(=O)=O

InChI

1S/C8H9NO4S/c1-13-8(10)6-4-2-3-5-7(6)14(9,11)12/h2-5H,1H3,(H2,9,11,12)

InChI key

VSOOBQALJVLTBH-UHFFFAOYSA-N

Gene Information

Application

Methyl 2-(aminosulfonyl)benzoate was used in the preparation of pyrazol-benzenesulfonamides, 2-(1-phenyl-1H-pyrazol-5-yl)benzenesulfonamides[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of 2-(1-Phenyl-1H-pyrazol-5-yl) benzenesulfonamides from Polylithiated C (alpha), N-Phenylhydrazones and Methyl 2-(Aminosulfonyl) benzoate.
Journal of Heterocyclic Chemistry, 42(6), 1095-1095 (2005)
P Serra-Mora et al.
The Science of the total environment, 721, 137732-137732 (2020-03-17)
The degradation kinetics and residual levels of the sulfonylurea herbicide tribenuron-methyl (TBM) in different environmental waters were studied using in tube-solid phase microextraction (IT-SPME) coupled on-line to nano-liquid chromatography (nanoLC) and UV diode array detection (DAD). This approach combines the
Rajae Chahboune et al.
Rapid communications in mass spectrometry : RCM, 29(15), 1370-1380 (2015-07-07)
Sulfonylureas are among the most important class of antidiabetic and herbicides. Solar light excitation and Advanced Oxidation Processes may result in the formation of a wide array of products owing to the relative complex structure. These products, that should be

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service