Skip to Content
Merck
All Photos(2)

Key Documents

244554

Sigma-Aldrich

Barium chlorate monohydrate

98%

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$104.00

$104.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 G
$104.00

About This Item

Linear Formula:
Ba(ClO3)2·H2O
CAS Number:
Molecular Weight:
322.24
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22
grade:
for analytical purposes

$104.00


Check Cart for Availability

Request a Bulk Order

grade

for analytical purposes

Quality Level

Assay

98%

mp

414 °C (lit.)

density

3.18 g/mL at 25 °C (lit.)

SMILES string

O.[Ba++].[O-]Cl(=O)=O.[O-]Cl(=O)=O

InChI

1S/Ba.2ClHO3.H2O/c;2*2-1(3)4;/h;2*(H,2,3,4);1H2/q+2;;;/p-2

InChI key

CIADYQPMTWAHHH-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Other Notes

Hydration may vary slightly

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Ox. Sol. 1

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service