1.01738
Barium perchlorate
anhydrous for analysis EMSURE®
Synonym(s):
Barium perchlorate
About This Item
Recommended Products
Quality Level
product line
EMSURE®
Assay
≥97.0% (complexometric)
form
solid
impurities
≤0.2% Substances not precipitated by sulfuric acid (as sulfates)
≤4% Water
pH
5.6 (20 °C, 100 g/L in H2O)
mp
505 °C
solubility
1985 g/L
density
2.74 g/cm3 at 20 °C
bulk density
1300 kg/m3
anion traces
chloride (Cl-): ≤0.01%
cation traces
Ca: ≤0.05%
Fe: ≤0.0005%
heavy metals (as Pb): ≤0.0005%
storage temp.
2-30°C
SMILES string
[Ba+2].[Cl](=O)(=O)(=O)[O-].[Cl](=O)(=O)(=O)[O-]
InChI
1S/Ba.2ClHO4/c;2*2-1(3,4)5/h;2*(H,2,3,4,5)/q+2;;/p-2
InChI key
OOULUYZFLXDWDQ-UHFFFAOYSA-L
Analysis Note
Chloride (Cl): ≤ 0.01 %
Heavy metals (as Pb): ≤ 0.0005 %
Ca (Calcium): ≤ 0.05 %
Fe (Iron): ≤ 0.0005 %
Water: ≤ 4.0 %
Substances not precipitated by sulfuric acid (as sulfates): ≤ 0.20 %
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Ox. Sol. 1
Storage Class Code
5.1A - Strongly oxidizing hazardous materials
WGK
WGK 1
Flash Point(F)
69.8 °F - closed cup
Flash Point(C)
21 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service