Skip to Content
Merck
All Photos(1)

Key Documents

240508

Sigma-Aldrich

1,1,2-Trichlorotrifluoroethane

anhydrous, 99.8%

Synonym(s):

1,1,2-Trichloro-1,2,2-trifluoroethane, 1,1,2-Trichloro-trifluoroethane, Genetron® 113

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCF2CCl2F
CAS Number:
Molecular Weight:
187.38
Beilstein:
1740335
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.21

grade

anhydrous

vapor density

2.39 (vs air)

vapor pressure

18.81 psi ( 55 °C)
5.5 psi ( 20 °C)
5.69 psi ( 20 °C)

Assay

99.8%

form

liquid

autoignition temp.

1256 °F

impurities

<0.002% water
<0.005% water (100 mL pkg)

evapn. residue

<0.0005%

refractive index

n20/D 1.358 (lit.)

bp

47-48 °C (lit.)

mp

−35 °C (lit.)

density

1.57 g/mL at 25 °C (lit.)

SMILES string

FC(F)(Cl)C(F)(Cl)Cl

InChI

1S/C2Cl3F3/c3-1(4,6)2(5,7)8

InChI key

AJDIZQLSFPQPEY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Packaged under nitrogen in Sure/Seal bottles

Legal Information

Genetron is a registered trademark of Honeywell International Inc.
Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Ozone 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

383.0 °F

Flash Point(C)

195 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mette Christophersen et al.
Journal of contaminant hydrology, 81(1-4), 1-33 (2005-08-17)
An emplaced hydrocarbon source field experiment was conducted in the relatively homogeneous sandy geology of the vadose zone at Airbase Vaerløse, Denmark. The source (10.2 l of NAPL) consisted of 13 hydrocarbons (n-, iso- and cyclo-alkanes and aromates) and CFC-113
H K Xiao et al.
American Industrial Hygiene Association journal, 51(7), 402-404 (1990-07-01)
Results obtained using Fourier transform infrared spectrophotometry (FTIR) for the analysis of samples of carbon disulfide (CS2) eluates containing trichloroethylene (TCE) and Freon from charcoal air sampling tubes were evaluated by comparison with results obtained when using gas chromatography (GC).
J A Richt et al.
Medical microbiology and immunology, 182(5), 271-280 (1993-11-01)
Treatment of homogenates from Borna disease virus (BDV)-infected brain tissue or cell cultures with Freon-113 yielded infectious particles with a buoyant density of 1.16-1.22 g/ml. Positive- and negative-stranded BDV-specific RNA species as well as three virus-specific proteins, known to be
Y Michael et al.
International journal of pharmaceutics, 221(1-2), 165-174 (2001-06-09)
The nature of the drug-drug aggregation phenomena between salmeterol xinafoate and fluticasone propionate used in a metered-dose inhaler system has been examined. Interactions between the drugs in the solvents 1,1,2-trichlorotrifloroethane (CFC-113) and 1,1,1,2-tetrafluoroethane (HFA-134a) have been characterised using a focused
S Sousa et al.
The Journal of applied bacteriology, 63(6), 559-564 (1987-12-01)
An initial investigation was made of the bactericidal and sporicidal activity of the dry cleaning solvent Freon-113 (1,1,2-trichlorotrifluoroethane) under various conditions. Representative bacterial strains selected for this study were Escherichia coli (ATCC 11229), Staphylococcus aureus (ATCC 6538) and spores of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service