Merck
  • Analysis of virus-specific RNA species and proteins in Freon-113 preparations of the Borna disease virus.
Analysis of virus-specific RNA species and proteins in Freon-113 preparations of the Borna disease virus.
Medical microbiology and immunology (1993-11-01)
J A Richt, J E Clements, S Herzog, J Pyper, K Wahn, H Becht
ABSTRACT
Treatment of homogenates from Borna disease virus (BDV)-infected brain tissue or cell cultures with Freon-113 yielded infectious particles with a buoyant density of 1.16-1.22 g/ml. Positive- and negative-stranded BDV-specific RNA species as well as three virus-specific proteins, known to be present in BDV-infected cell extracts, were demonstrated in these Freon-treated fractions. When the Freon-purified virus preparations were treated with RNase A prior to RNA extraction, only negative-stranded, genomic RNA was detected in Northern blot hybridizations using sense and antisense RNA probes. These data substantiate that BDV is a negative-stranded RNA virus.