Skip to Content
Merck
All Photos(1)

Key Documents

232963

Sigma-Aldrich

4-(Trifluoromethyl)phenylacetonitrile

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$41.40

$41.40


Estimated to ship on01 April 2025


Request a Bulk Order

Select a Size

Change View
5 G
$41.40

About This Item

Linear Formula:
CF3C6H4CH2CN
CAS Number:
Molecular Weight:
185.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$41.40


Estimated to ship on01 April 2025


Request a Bulk Order

Quality Level

Assay

98%

form

solid

bp

131-132 °C/20 mmHg (lit.)

mp

47-49 °C (lit.)

functional group

fluoro
nitrile

SMILES string

FC(F)(F)c1ccc(CC#N)cc1

InChI

1S/C9H6F3N/c10-9(11,12)8-3-1-7(2-4-8)5-6-13/h1-4H,5H2

InChI key

QNKOCFJZJWOXDE-UHFFFAOYSA-N

Application

4-(Trifluoromethyl)phenylacetonitrile was used in the preparation of:
  • 1,4-bis[2-cyano-2-(4-(trifluoromethyl)phenyl)vinyl]benzene[1]
  • cyano-substituted distyrylbenzene derivative, novel n-type organic semiconductor[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Steady Operation of n-Type Organic Thin-Film Transistors with Cyano-Substituted Distyrylbenzene Derivative.
Nagamatsu S, et al.
Applied Physics Express, 2(10), 101502-101502 (2009)
Synthesis and FET characteristics of phenylene-vinylene and anthracene-vinylene compounds containing cyano groups.
Shoji K, et al.
Journal of Materials Chemistry, 20(31), 6472-6478 (2010)
Immacolata Serra et al.
Marine biotechnology (New York, N.Y.), 21(2), 229-239 (2019-01-27)
A screening among marine yeasts was carried out for nitrile hydrolyzing activity. Meyerozyma guilliermondii LM2 (UBOCC-A-214008) was able to efficiently grow on benzonitrile and cyclohexanecarbonitrile (CECN) as sole nitrogen sources. A two-step one-pot method for obtaining cells of M. guilliermondii

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service