Skip to Content
Merck
All Photos(1)

Key Documents

102407

Sigma-Aldrich

4-Fluorophenylacetonitrile

99%

Synonym(s):

4-Fluorobenzyl cyanide

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$116.00

$116.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$116.00

About This Item

Linear Formula:
FC6H4CH2CN
CAS Number:
Molecular Weight:
135.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$116.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

refractive index

n20/D 1.5002 (lit.)

bp

119-120 °C/18 mmHg (lit.)

density

1.126 g/mL at 25 °C (lit.)

functional group

fluoro
nitrile

SMILES string

Fc1ccc(CC#N)cc1

InChI

1S/C8H6FN/c9-8-3-1-7(2-4-8)5-6-10/h1-4H,5H2

InChI key

JHQBLYITVCBGTO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Fluorophenylacetonitrile is starting reagent in the synthesis of 1-Alkyl-N-[2-ethyl-2-(4-fluorophenyl)butyl]piperidine-4-carboxamide derivatives[1].

Biochem/physiol Actions

4-Fluorophenylacetonitrile undergoes biotransformation to 4-Fluorophenylacetic acid by marine fungi, Aspergillus sydowii Ce19[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

226.4 °F - closed cup

Flash Point(C)

108 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Susumu Watanuki et al.
Bioorganic & medicinal chemistry, 19(18), 5628-5638 (2011-08-31)
We synthesized and evaluated inhibitory activity against T-type Ca(2+) channels for a series of 1-alkyl-N-[2-ethyl-2-(4-fluorophenyl)butyl]piperidine-4-carboxamide derivatives. Structure-activity relationship studies have revealed that dialkyl substituents at the benzylic position play an important role in increasing inhibitory activity. Oral administration of N-[2-ethyl-2-(4-fluorophenyl)butyl]-1-(2-phenylethyl)piperidine-4-carboxamide
Julieta Rangel de Oliveira et al.
Marine biotechnology (New York, N.Y.), 15(1), 97-103 (2012-07-14)
Marine fungi belonging to the genera Aspergillus, Penicillium, Cladosporium, and Bionectria catalyzed the biotransformation of phenylacetonitrile to 2-hydroxyphenylacetic acid. Eight marine fungi, selected and cultured with phenylacetonitrile in liquid mineral medium, catalyzed it quantitative biotransformation to 2-hydroxyphenylacetic acid. In this

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service