Skip to Content
Merck
All Photos(2)

Key Documents

224502

Sigma-Aldrich

Methyl 4-hydroxyphenylacetate

ReagentPlus®, 99%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$108.00
100 G
$254.00

$108.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$108.00
100 G
$254.00

About This Item

Linear Formula:
HOC6H4CH2CO2CH3
CAS Number:
Molecular Weight:
166.17
Beilstein:
2087538
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$108.00


Check Cart for Availability

Request a Bulk Order

Quality Level

product line

ReagentPlus®

Assay

99%

form

solid

bp

162-163 °C/5 mmHg (lit.)

mp

55-58 °C (lit.)

functional group

ester

SMILES string

COC(=O)Cc1ccc(O)cc1

InChI

1S/C9H10O3/c1-12-9(11)6-7-2-4-8(10)5-3-7/h2-5,10H,6H2,1H3

InChI key

XGDZEDRBLVIUMX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Methyl 4-hydroxyphenylacetate inhibits the activity of tobacco mosaic virus (TMV)[1].

Application

Methyl 4-hydroxyphenylacetate was used in the synthesis of 4-(2-hydroxy-3-isopropylamino)propoxyphenylacetic acid[2].

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuo Shen et al.
Natural product research, 27(24), 2286-2291 (2013-08-22)
A new compound 2-(4-hydroxybenzoyl) quinazolin-4(3H)-one (1) and four known compounds were isolated from a marine fungus Penicillium oxalicum 0312F1. The structure of the new compound 1 was elucidated based on the spectroscopic analysis. Bioactivity assays showed that 2-(4-hydroxybenzyl) quinazolin-4(3H)-one (2)
S H Jung et al.
Archives of pharmacal research, 23(3), 226-229 (2000-07-15)
To obtain the standard compounds of metoprolol for a pharmacokinetic study, a convenient synthetic procedure to prepare enantiomers of metoprolol (3a) and its major metabolites, 2-4-(2-hydroxy-3-isopropylamino)propoxyphenylethanol (3b) and 4-(2-hydroxy-3-isopropylamino)propoxyphenylacetic acid (4), was developed from their respective starting materials, 4-(2-methoxyethyl)phenol (1a)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service