Skip to Content
Merck
All Photos(1)

Documents

72813

Sigma-Aldrich

Oenin chloride

≥90% (HPLC)

Synonym(s):

Cyclamin chloride, Enin chloride, Malvidin 3-β-D-glucopyranoside, Malvidin-3-O-glucoside chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H25ClO12
CAS Number:
Molecular Weight:
528.89
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

Assay

≥90% (HPLC)

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

SMILES string

[Cl-].COc1cc(cc(OC)c1O)-c2[o+]c3cc(O)cc(O)c3cc2O[C@@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O

InChI

1S/C23H24O12.ClH/c1-31-14-3-9(4-15(32-2)18(14)27)22-16(7-11-12(26)5-10(25)6-13(11)33-22)34-23-21(30)20(29)19(28)17(8-24)35-23;/h3-7,17,19-21,23-24,28-30H,8H2,1-2H3,(H2-,25,26,27);1H/t17-,19-,20+,21-,23-;/m1./s1

InChI key

YDIKCZBMBPOGFT-DIONPBRTSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Oenin chloride/malvidin-3-O-glucoside is an anthocyanin content, present at high level in Vitis vinifera young red wines. It helps in defining the colour of wine.

Biochem/physiol Actions

Anthocyanin. Studied for its neuroprotective properties.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oenin and Quercetin Copigmentation: Highlights From Density Functional Theory
Li Y, et al.
Frontiers in Chemistry, 6, 245-245 (2018)
Stefanos Koundouras et al.
Journal of agricultural and food chemistry, 57(17), 7805-7813 (2009-09-03)
Compositional changes of skin and seed phenolic compounds and berry glycosylated aroma precursors were measured in Vitis vinifera L. cv. Cabernet Sauvignon onto 1103P and SO4 rootstocks, in three irrigation regimes (FI, 100% of evapotranspiration; DI, 50% of evapotranspiration; and
Iva Fernandes et al.
Food & function, 3(5), 508-516 (2012-03-07)
The bioavailability of anthocyanins is the most difficult one to assess amongst all flavonoid compounds as a result of their occurrence under different structures in equilibrium depending on pH. Due to their rapid appearance in plasma, the absorption of anthocyanins
Baoshan Sun et al.
Journal of agricultural and food chemistry, 57(18), 8623-8627 (2009-09-01)
Phenolic complexes are a major group of polyphenols in aged red wine. The objective of this work was to evaluate the in vitro antioxidant activity of the phenolic complexes. Thus, red wine polyphenols were fractionated into various fractions including monomers
Luís Cruz et al.
Rapid communications in mass spectrometry : RCM, 26(18), 2123-2130 (2012-08-14)
The aim of this work was to study the putative reactions that could occur in red wines between malvidin-3-glucoside-(O)-catechin (mv3glc-(O)-cat) adduct and catechin (cat) or malvidin-3-glucoside (mv3glc) in presence of acetaldehyde. Mv3glc-(O)-cat adduct (1 mM) was incubated with catechin (or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service