Skip to Content
Merck
All Photos(2)

Key Documents

210080

Sigma-Aldrich

N-Methylaniline trifluoroacetate

99%

Synonym(s):

TAMA, Trifluoroacetic acid N-methylaniline salt

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$69.20

$69.20


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$69.20

About This Item

Linear Formula:
C6H5NH2+(CH3)CF3COO-
CAS Number:
Molecular Weight:
221.18
Beilstein:
3638133
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$69.20


Check Cart for Availability

Request a Bulk Order

Assay

99%

form

solid

mp

65-66 °C (lit.)

functional group

amine
carboxylic acid
fluoro

SMILES string

OC(=O)C(F)(F)F.CNc1ccccc1

InChI

1S/C7H9N.C2HF3O2/c1-8-7-5-3-2-4-6-7;3-2(4,5)1(6)7/h2-6,8H,1H3;(H,6,7)

InChI key

TXXJGCVOHDSYBC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenjian Qian et al.
Bioorganic & medicinal chemistry, 21(14), 3996-4003 (2012-06-30)
Protein-protein interactions (PPIs) mediated by the polo-box domain (PBD) of polo-like kinase 1 (Plk1) serve important roles in cell proliferation. Critical elements in the high affinity recognition of peptides and proteins by PBD are derived from pThr/pSer-residues in the binding
Annefleur E M Berkel et al.
Digestive surgery, 36(5), 376-383 (2018-06-14)
To investigate the relation between skeletal muscle measurements (muscle mass, radiation attenuation, and sarcopenic obesity), postoperative morbidity, and survival after treatment of locally advanced rectal cancer. This explorative retrospective study identified 99 consecutive patients who underwent neoadjuvant chemoradiation and surgery
Synthetic studies on bradykinin antagonist martinellines: construction of a pyrrolo [3, 2-c] quinoline skeleton using silicon-tether RCM reaction and allylic amination.
Hara O, et al.
Tetrahedron, 60(42), 9381-9390 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service