N-Methylaniline trifluoroacetate
Synonym(s):
TAMA, Trifluoroacetic acid N-methylaniline salt
Linear Formula:
C6H5NH2+(CH3)CF3COO-
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing