Skip to Content
Merck
All Photos(1)

Key Documents

20787

Sigma-Aldrich

1D-1-O-Butyryl-4,6-O-dibenzoyl-myo-inositol

≥98.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H26O9
CAS Number:
Molecular Weight:
458.46
Beilstein:
6620437
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0%

SMILES string

CCCC(=O)O[C@@H]1[C@H](O)[C@H](O)[C@@H](OC(=O)c2ccccc2)[C@H](O)[C@H]1OC(=O)c3ccccc3

InChI

1S/C24H26O9/c1-2-9-16(25)31-21-18(27)17(26)20(32-23(29)14-10-5-3-6-11-14)19(28)22(21)33-24(30)15-12-7-4-8-13-15/h3-8,10-13,17-22,26-28H,2,9H2,1H3/t17-,18+,19-,20+,21+,22+/m0/s1

InChI key

KTYAOQKEMRXFIY-NXOOKBFXSA-N


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael A Wheeler et al.
Cell, 176(3), 581-596 (2019-01-22)
Genome-wide studies have identified genetic variants linked to neurologic diseases. Environmental factors also play important roles, but no methods are available for their comprehensive investigation. We developed an approach that combines genomic data, screens in a novel zebrafish model, computational
Michael A Wheeler et al.
Nature, 578(7796), 593-599 (2020-02-14)
Multiple sclerosis is a chronic inflammatory disease of the CNS1. Astrocytes contribute to the pathogenesis of multiple sclerosis2, but little is known about the heterogeneity of astrocytes and its regulation. Here we report the analysis of astrocytes in multiple sclerosis
Chun-Cheih Chao et al.
Cell, 179(7), 1483-1498 (2019-12-10)
Metabolism has been shown to control peripheral immunity, but little is known about its role in central nervous system (CNS) inflammation. Through a combination of proteomic, metabolomic, transcriptomic, and perturbation studies, we found that sphingolipid metabolism in astrocytes triggers the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service