Skip to Content
Merck
All Photos(1)

Key Documents

20589

Sigma-Aldrich

O-tert-Butyl-L-serine tert-butyl ester hydrochloride

≥98.0% (AT)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H23NO3 · HCl
CAS Number:
Molecular Weight:
253.77
Beilstein:
4886392
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (AT)

form

solid

optical activity

[α]/D -10.0±1.0°, c = 1 in H2O

reaction suitability

reaction type: solution phase peptide synthesis

application(s)

peptide synthesis

SMILES string

Cl.CC(C)(C)OC[C@H](N)C(=O)OC(C)(C)C

InChI

1S/C11H23NO3.ClH/c1-10(2,3)14-7-8(12)9(13)15-11(4,5)6;/h8H,7,12H2,1-6H3;1H/t8-;/m0./s1

InChI key

RDWZQVGVBTYCBD-QRPNPIFTSA-N

Other Notes

Protected L-serine used in peptide synthesis, e.g. synthesis of glycopeptides and lipopeptides[1][2][3]; Preparation of amino-phospholipids[4]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liebigs Ann. Chem., 771-771 (1989)
J G Turcotte et al.
Chemistry and physics of lipids, 58(1-2), 81-95 (1991-05-01)
A homologous series of chiral (R) ether-amide phosphonolipid analogs of naturally occurring (R) glycerophospholipids were synthesized and characterized for their interfacial behaviors. The phosphonolipids possess isoteric ether, amide, and phosphonate functions at positions corresponding to the sn-1, sn-2, and sn-3
Y Tsuda et al.
Chemical & pharmaceutical bulletin, 39(3), 607-611 (1991-03-01)
The structure of WS1279, isolated from Streptomyces sp. as an immunoactive lipopeptide, has been deduced on the basis of chemical and physical evidence as S-[2,3-bis(palmitoyloxy)propyl]-N alpha-palmitoyl-Cys-Asn-Ser-Gly-Gly-Ser- OH. This was confirmed by synthesis.
H. Paulsen et al.
Liebigs Ann. Chem., 751-751 (1989)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service