Merck
  • Chemical synthesis and surface activity of lung surfactant phospholipid analogs. III. Chiral N-substituted ether-amide phosphonolipids.
Chemical synthesis and surface activity of lung surfactant phospholipid analogs. III. Chiral N-substituted ether-amide phosphonolipids.
Chemistry and physics of lipids (1991-05-01)
J G Turcotte, W H Lin, N C Motola, P E Pivarnik, N N Bhongle, H R Heyman, S S Shirali, Z Lu, R H Notter
ABSTRACT
A homologous series of chiral (R) ether-amide phosphonolipid analogs of naturally occurring (R) glycerophospholipids were synthesized and characterized for their interfacial behaviors. The phosphonolipids possess isoteric ether, amide, and phosphonate functions at positions corresponding to the sn-1, sn-2, and sn-3 ester functions, respectively, of naturally occurring glycerophospholipids. All compounds were synthesized with disaturated C16:0 alkyl/acyl moieties to give structural analogy with dipalmitoyl phosphatidylcholine (DPPC), the major glycerophospholipid component of lung surfactant. Further substitutions at the headgroup nitrogen were also used to generate differences in headgroup size and polarity in the synthetic compounds. The surface activity of the ether-amide phospholipids was investigated in terms of adsorption to the air-water interface, together with studies of dynamic respreading after monolayer collapse and surface tension lowering in dynamically compressed spread films and dispersions. Results showed that several ether-amide phosphonolipids had more rapid adsorption and improved dynamic respreading behavior compared to DPPC, plus the ability to lower surface tension into the range of less than 1 to 4 mN/m in spread films and in dispersions under dynamic conditions. In combination with a series of diether phosphonolipids synthetized in a companion study [1], these ether-amide compounds are useful in the development of molecular structure-surface activity correlates for lung surfactant-related materials, and should assist in investigating the specificity of interactions between phospholipids and other pulmonary biological molecules.