Skip to Content
Merck
All Photos(1)

Key Documents

205206

Sigma-Aldrich

2-Amino-5-bromo-6-methyl-4-pyrimidinol

97%

Synonym(s):

5-Bromo-6-methylisocytosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6BrN3O
CAS Number:
Molecular Weight:
204.02
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

244-246 °C (lit.)

functional group

bromo

SMILES string

Cc1nc(N)nc(O)c1Br

InChI

1S/C5H6BrN3O/c1-2-3(6)4(10)9-5(7)8-2/h1H3,(H3,7,8,9,10)

InChI key

ADLWOFHKMXUDKF-UHFFFAOYSA-N

General description

2-Amino-5-bromo-6-methyl-4-pyrimidinol induced high levels of serum interferon in mice[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P S Morahan et al.
Antiviral research, 15(3), 241-254 (1991-03-01)
The question of whether interferon alpha/beta is the common mechanism of antiviral action of synthetic immunomodulators was investigated in B6C3F1 mice infected with Semliki Forest virus. Mice were treated with various concentrations of normal sheep serum or potent anti-alpha/beta interferon
L S Anthony et al.
Clinical and investigative medicine. Medecine clinique et experimentale, 7(4), 343-348 (1984-01-01)
The modulation of murine host resistance to infection with Listeria monocytogenes by the substituted pyrimidine anti-viral compounds, 2-amino-5-bromo-6-methyl-4-pyrimidinol (ABMP), 2-amino-5-bromo-6-phenyl-4-pyrimidinol (ABPP) and 2-amino-5-iodo-6-phenyl-4-pyrimidinol (AIPP) was investigated. BAF1 mice given three daily injections of ABMP, ABPP (as well as of the
Antineoplastic properties of pyrimidinone interferon inducers.
D A Stringfellow
Advances in enzyme regulation, 19, 335-348 (1980-01-01)
D A Stringfellow et al.
Journal of interferon research, 1(1), 1-14 (1980-01-01)
The interferon inducing characteristics of a new series of 6-phenyl pyrimidinol compounds are described and compared against a previously identified pyrimidine, 2-amino-5-bromo-6-methyl-4-pyrimidinol (ABMP). Interestingly, a split in ability to induce interferon but not in vivo antiviral activity was observed in
D A Stringfellow
Antimicrobial agents and chemotherapy, 17(3), 455-460 (1980-03-01)
Prostaglandins enhanced the interferon response and therapeutic activity of interferon inducers in Friend leukemia virus-infected mice. A similar enhancement in interfron responsiveness but not antiviral activity was observed in mice infected with rapidly acute viruses.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service