Recommended Products
Quality Level
Assay
98%
mp
198-201 °C (lit.)
SMILES string
OC(=O)c1ccccc1C(=O)c2ccc(Cl)c(c2)[N+]([O-])=O
InChI
1S/C14H8ClNO5/c15-11-6-5-8(7-12(11)16(20)21)13(17)9-3-1-2-4-10(9)14(18)19/h1-7H,(H,18,19)
InChI key
RITAQDHCJBLSSL-UHFFFAOYSA-N
General description
Reduction of 2-(4-chloro-3-nitrobenzoyl)benzoic acid in 5% NaOH aqueous solution in presence of Raney nickel, Pd/Al2O3 and Pt/Al2O3 has been investigated.
Application
2-(4-Chloro-3-nitrobenzoyl)benzoic acid has been used in the preparation of indanthrone derivatives.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
475. Chemistry of indanthrone. Part VI. Alkylsulphonyl-indanthrones.
Journal of the Chemical Society, 2177-2185 (1951)
Catalytic Liquid-Phase Reduction of Aromatic Nitro Compounds Containing Highly Reactive Functional Groups.
Russ. J. Org. Chem., 37(5), 673-676 (2001)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service