Skip to Content
Merck
All Photos(1)

Key Documents

530913

Sigma-Aldrich

3-Butoxypropionitrile

98%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$86.70

$86.70


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$86.70

About This Item

Linear Formula:
CH3(CH2)3O(CH2)2CN
CAS Number:
Molecular Weight:
127.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$86.70


Check Cart for Availability

Request a Bulk Order

Assay

98%

refractive index

n20/D 1.421 (lit.)

bp

210 °C (lit.)

density

0.925 g/mL at 25 °C (lit.)

functional group

ether
nitrile

SMILES string

CCCCOCCC#N

InChI

1S/C7H13NO/c1-2-3-6-9-7-4-5-8/h2-4,6-7H2,1H3

InChI key

AWVNJBFNHGQUQU-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

192.9 °F - closed cup

Flash Point(C)

89.4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Understanding and Successfully Applying Materials for Dye-sensitized solar cells.
Desilvestro H, et al.
Material Matters, 9(1), 14-17 (2014)
Amido-imidol tautomerization by acid-catalyzed addition of nitriles to 16-hydroxyhexadec-cis-9-enoic acid: A novel route for 9-[substituted amido]-16-ol-hexadecanoic acids, and their biological importance and possible industrial utilization.
Hosamani KM and Sattigeri RM.
Industrial & Engineering Chemistry Fundamentals, 44(2), 254-260 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service