Skip to Content
Merck
All Photos(2)

Key Documents

195367

Sigma-Aldrich

2-Methyl-3-nitrobenzyl chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NO2)CH2Cl
CAS Number:
Molecular Weight:
185.61
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

43-46 °C (lit.)

SMILES string

Cc1c(CCl)cccc1[N+]([O-])=O

InChI

1S/C8H8ClNO2/c1-6-7(5-9)3-2-4-8(6)10(11)12/h2-4H,5H2,1H3

InChI key

XZNDXQGZPOZITR-UHFFFAOYSA-N

General description

2-Methyl-3-nitrobenzyl chloride is a lachrymatory agent[1].

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Review of US Patents in the Field of Organic Process Development Published During October and November 2009.
Turner K.
Organic Process Research & Development, 14(2), 311-322 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service