Skip to Content
Merck
All Photos(1)

Key Documents

275530

Sigma-Aldrich

(2-Pyrimidylthio)acetic acid

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$62.40

$62.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$62.40

About This Item

Empirical Formula (Hill Notation):
C6H6N2O2S
CAS Number:
Molecular Weight:
170.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$62.40


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

powder

mp

204 °C (dec.) (lit.)

functional group

carboxylic acid
thioether

SMILES string

OC(=O)CSc1ncccn1

InChI

1S/C6H6N2O2S/c9-5(10)4-11-6-7-2-1-3-8-6/h1-3H,4H2,(H,9,10)

InChI key

NIEOYUNNKKAQKI-UHFFFAOYSA-N

Application

(2-Pyrimidylthio)acetic acid was used in the synthesis of diorganotin(IV)-oxo-carboxylates by reacting with diorganotin(IV) oxide[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Syntheses, spectroscopic characterizations and crystal structures of diorganotin (IV)-oxo-carboxylates with 2-pyrazinecarboxylic acid or (2-pyrimidylthio) acetic acid.
Journal of Coordination Chemistry, 59(12), 1335-1347 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service