Skip to Content
Merck
All Photos(1)

Documents

194352

Sigma-Aldrich

Ethyl 1-methyl-3-piperidinecarboxylate

96%

Synonym(s):

1-Methyl-nipecotic acid ethyl ester, Ethyl 1-methylnipecotate, NSC 64739

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H17NO2
CAS Number:
Molecular Weight:
171.24
Beilstein:
123259
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

refractive index

n20/D 1.451 (lit.)

bp

88-89 °C/11 mmHg (lit.)

density

0.954 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)C1CCCN(C)C1

InChI

1S/C9H17NO2/c1-3-12-9(11)8-5-4-6-10(2)7-8/h8H,3-7H2,1-2H3

InChI key

VFJJNMLPRDRTCO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Enantiomeric discrimination has been investigated in 1H and 13C NMR spectra of ethyl 1-methyl-3-piperidinecarboxylate in the presence of (-)-(18-crown-6)-2,3,11,12-tetracarboxylic acid.

Application

Reactant for:
N-demethylation of tertiary amines
Reverse cope elimination reactions
Regioselective oxidation
Synthesis of chelating agents
Sequential Michael-Michael-Dieckmann cyclizations

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

154.4 °F - closed cup

Flash Point(C)

68 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ming-Hsui Tsai et al.
Psychiatry research, 271, 153-160 (2018-11-27)
The purposes of this study were to develop the Chinese-version betel quid dependence instrument (BQDI) and to test its reliability and validity. An item pool relevant to betel quid dependence was generated. A panel of three experts assessed content validity
Ann E Lovely et al.
The Journal of organic chemistry, 71(24), 9178-9182 (2006-11-18)
Enantiomeric discrimination is observed in the (1)H and (13)C NMR spectra of piperidines and piperazines in the presence of (-)-(18-crown-6)-2,3,11,12-tetracarboxylic acid. The amines are protonated by the carboxylic acid groups of the crown ether to produce the corresponding ammonium and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service