Skip to Content
Merck
All Photos(1)

Key Documents

192406

Sigma-Aldrich

2-Chloropropionitrile

95%

Synonym(s):

α-Chloropropionitrile, (±)-2-Chloropropionitrile, 2-Chloropropanenitrile

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$189.00
25 G
$791.00

$189.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$189.00
25 G
$791.00

About This Item

Linear Formula:
CH3CH(Cl)CN
CAS Number:
Molecular Weight:
89.52
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$189.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

liquid

refractive index

n20/D 1.413 (lit.)

bp

120-122 °C (lit.)

density

1.012 g/mL at 25 °C (lit.)

functional group

chloro
nitrile

SMILES string

CC(Cl)C#N

InChI

1S/C3H4ClN/c1-3(4)2-5/h3H,1H3

InChI key

JNAYPRPPXRWGQO-UHFFFAOYSA-N

Application

2-Chloropropionitrile was used as initiator during the single-pot atom-transfer radical polymerization under microwave irradiation for the synthesis of polyacrylonitrile[1]. It was used in the synthesis of:
  • nitrile functionalized methimazole-based ionic liquids[2]
  • (+)- and (-)-anatoxin[3]

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

91.4 °F - closed cup

Flash Point(C)

33 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ATRP of acrylonitrile catalyzed by FeCl2/succinic acid under microwave irradiation.
Hou C, et al.
Journal of Applied Polymer Science, 101(3), 1598-1601 (2006)
Efficient new syntheses of (+)-and (-)-anatoxin-a. Revised configuration of resolved 9-methyl-9-azabicyclo [4.2. 1] nonan-2-one.
Ferguson JR, et al.
Tetrahedron Letters, 36(48), 8867-8870 (1995)
Amal I Siriwardana et al.
The Journal of organic chemistry, 75(24), 8376-8382 (2010-11-18)
The alkylation reaction of 2-mercapto-1-methylimidazole 1b with 2-chloroacetonitrile and 2-chloropropionitrile produced S-alkyl methimazole chlorides 2a and 2b which were subjected to anion metathesis with lithium bis(trifluoromethanesulfonyl)amide, LiNTf(2), to afford nitrile functionalized methimazole-based room temperature ionic liquids 3a and 3b in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service