Skip to Content
Merck
All Photos(1)

Key Documents

560626

Sigma-Aldrich

2-Isopropenyl-2-oxazoline

98%

Synonym(s):

2-Isopropenyl-2-oxazoline

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$360.00

$360.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$360.00

About This Item

Empirical Formula (Hill Notation):
C6H9NO
CAS Number:
Molecular Weight:
111.14
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$360.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

contains

1000 ppm phenothiazine as inhibitor

refractive index

n20/D 1.478 (lit.)

viscosity

0.993 cP(25 °C)

bp

147 °C (lit.)

mp

−47 °C (lit.)

density

1.06 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(=C)C1=NCCO1

InChI

1S/C6H9NO/c1-5(2)6-7-3-4-8-6/h1,3-4H2,2H3

InChI key

LPIQIQPLUVLISR-UHFFFAOYSA-N

General description

Soluble in water and organic solvents

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

118.4 °F - closed cup

Flash Point(C)

48 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michał Cegłowski et al.
Pharmaceutics, 12(6) (2020-06-06)
Trigger-responsive materials are capable of controlled drug release in the presence of a specific trigger. Reduction induced drug release is especially interesting as the reductive stress is higher inside cells than in the bloodstream, providing a conceptual controlled release mechanism
Joachim F R Van Guyse et al.
ACS applied materials & interfaces, 11(34), 31356-31366 (2019-08-06)
Plasma polymerization is gaining popularity as a technique for coating surfaces due to the low cost, ease of operation, and substrate-independent nature. Recently, the plasma polymerization (or deposition) of 2-oxazoline monomers was reported resulting in coatings that have potential applications
Tom Flossmann et al.
Cell reports, 26(12), 3173-3182 (2019-03-21)
Synchronized activity is a universal characteristic of immature neural circuits that is essential for their developmental refinement and strongly depends on GABAergic neurotransmission. A major subpopulation of GABA-releasing interneurons (INs) expresses somatostatin (SOM) and proved critical for rhythm generation in
Arthur H Cheng et al.
Cell reports, 26(12), 3191-3202 (2019-03-21)
Clock neurons within the mammalian suprachiasmatic nuclei (SCN) encode circadian time using interlocked transcription-translation feedback loops (TTFLs) that drive rhythmic gene expression. However, the contributions of other transcription factors outside of the circadian TTFLs to the functionality of the SCN

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service