Skip to Content
Merck
All Photos(1)

Key Documents

191353

Sigma-Aldrich

5-Chloro-2-(methylamino)benzophenone

99%

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$79.60

$79.60


Available to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
10 G
$79.60

About This Item

Linear Formula:
CH3NHC6H3(Cl)COC6H5
CAS Number:
Molecular Weight:
245.70
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$79.60


Available to ship on22 April 2025Details


Request a Bulk Order

Quality Level

Assay

99%

form

solid

mp

93-95 °C (lit.)

SMILES string

CNc1ccc(Cl)cc1C(=O)c2ccccc2

InChI

1S/C14H12ClNO/c1-16-13-8-7-11(15)9-12(13)14(17)10-5-3-2-4-6-10/h2-9,16H,1H3

InChI key

WPNMLCMTDCANOZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

5-Chloro-2-(methylamino)benzophenone was used in the simultaneous determination of hydrazine and benzylhydrazine in isocarboxazid raw material and tablet formulations by gas chromatography[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Sajan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 106, 253-261 (2013-02-16)
In this paper, the vibrational spectral analysis and three-photon absorption properties of an organic material of 2-methylamino-5-chlorobenzophenone have been reported. The geometry and harmonic vibrational wavenumbers are calculated with the help of B3LYP density functional theory method. The detailed interpretation
M I Krut'
Sudebno-meditsinskaia ekspertiza, 41(3), 24-26 (1998-08-15)
Cibasone degradation products were measured in corpses of 15 cats poisoned with cibasone in a dose of 750 mg/kg. Cibasone was assessed by semiquantitative method on Silufol UV-254 plates from the content of the main degradation product 3-methylamino-5-chloro-benzophenone (MCB) in
N S Nudelman et al.
Journal of pharmaceutical sciences, 84(8), 998-1004 (1995-08-01)
In the acid hydrolysis of diazepam (1), several unusual products, apart from 2-(N-methylamino)-5-chlorobenzophenone (2) and glycine, were isolated. On the assumption that some of those products could arise from further degradation of 2, the reaction of this compound with 0.5-2
T Suthan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 79(5), 1443-1448 (2011-05-24)
The organic material 2-methylamino-5-chlorobenzophenone single crystal has been grown by modified vertical Bridgman technique using the single wall ampoule with nano translation. The grown crystal was confirmed by single crystal and powder X-ray diffraction analyses. Fourier transform infrared analysis was
R Jain
Journal of chromatography, 615(2), 365-368 (1993-06-02)
A direct densitometric method for determination of diazepam and its metabolites in urine was developed. The proposed procedure involves acid hydrolysis of urine specimens, thereby converting diazepam and its metabolites into benzophenones [2-methylamino-5-chlorobenzophenone (MACB) and 2-amino-5-chlorobenzophenone (ACB)]. It is followed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service