Skip to Content
Merck
All Photos(3)

Key Documents

B9602

Sigma-Aldrich

Benzophenone hydrazone

96%

Synonym(s):

Diphenyldiazomethane precursor

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$119.00
500 G
$456.00

$119.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$119.00
500 G
$456.00

About This Item

Linear Formula:
(C6H5)2C=NNH2
CAS Number:
Molecular Weight:
196.25
Beilstein:
1910177
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$119.00


Check Cart for Availability

Request a Bulk Order

Assay

96%

form

crystals

bp

225-230 °C/55 mmHg (lit.)

mp

95-98 °C (lit.)

storage temp.

2-8°C

SMILES string

N\N=C(/c1ccccc1)c2ccccc2

InChI

1S/C13H12N2/c14-15-13(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10H,14H2

InChI key

QYCSNMDOZNUZIT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

294.8 °F

Flash Point(C)

146 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arshia et al.
Bioorganic & medicinal chemistry, 27(6), 1009-1022 (2019-02-11)
This study deals with the synthesis of benzophenone sulfonamides hybrids (1-31) and screening against urease enzyme in vitro. Studies showed that several synthetic compounds were found to have good urease enzyme inhibitory activity. Compounds 1 (N'-((4'-hydroxyphenyl)(phenyl)methylene)-4''-nitrobenzenesulfonohydrazide), 2 (N'-((4'-hydroxyphenyl)(phenyl)methylene)-3''-nitrobenzenesulfonohydrazide), 3 (N'-((4'-hydroxyphenyl)(phenyl)methylene)-4''-methoxybenzenesulfonohydrazide)
Fatemeh M Mir et al.
Journal of medicinal chemistry, 62(9), 4500-4525 (2019-04-02)
Peptide mimicry employing a combination of aza-amino acyl proline and indolizidinone residues has been used to develop allosteric modulators of the prostaglandin F2α receptor. The systematic study of the N-terminal phenylacetyl moiety and the conformation and side chain functions of
Kelvine Chignen Possi et al.
Journal of medicinal chemistry, 60(22), 9263-9274 (2017-10-14)
Azapeptide analogues of growth hormone releasing peptide-6 (GHRP-6) exhibit promising affinity, selectivity, and modulator activity on the cluster of differentiation 36 receptor (CD36). For example, [A
Ngoc-Duc Doan et al.
Journal of peptide science : an official publication of the European Peptide Society, 21(5), 387-391 (2014-11-18)
The solid-phase synthesis of azapeptides possessing a C-terminal aza-residue has been accomplished by a protocol featuring regioselective alkylation of benzhydrylidene-aza-glycinamide and illustrated by the syntheses of [aza-Lys(6)] growth-hormone-releasing peptide-6 analogs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service