Skip to Content
Merck
All Photos(1)

Key Documents

170925

Sigma-Aldrich

4-Nitrobenzenesulfonyl chloride

technical grade, 90%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$83.40

$83.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$83.40

About This Item

Linear Formula:
O2NC6H4SO2Cl
CAS Number:
Molecular Weight:
221.62
Beilstein:
746543
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$83.40


Check Cart for Availability

Request a Bulk Order

grade

technical grade

Quality Level

Assay

90%

mp

66-70 °C (lit.)

SMILES string

[O-][N+](=O)c1ccc(cc1)S(Cl)(=O)=O

InChI

1S/C6H4ClNO4S/c7-13(11,12)6-3-1-5(2-4-6)8(9)10/h1-4H

InChI key

JXRGUPLJCCDGKG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Nitrobenzenesulfonyl chloride was used in the synthesis of [Cu(bipy)2Cl](nbs) where bipy = 2,2′-bipyridine, nbs = 4-nitrobenzenesulfonate[1]. It was also used to determine estrogens in biological fluids[2].

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Repr. 2 - Skin Sens. 1A

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tatsuya Higashi et al.
Analytical and bioanalytical chemistry, 386(3), 658-665 (2006-04-13)
A practical procedure for determining estrogens in biological fluids has been studied using liquid chromatography-electron capture atmospheric pressure chemical ionization-mass spectrometry combined with derivatization. Among the commercially available reagents (4-nitrobenzoyl chloride, 2,4-dinitrofluorobenzene, 4-nitrobenzenesulfonyl chloride and 4-nitrobenzyl bromide), 4-nitrobenzenesulfonyl chloride was
Mahboubeh A Sharif et al.
Acta chimica Slovenica, 59(2), 289-293 (2012-06-01)
[Cu(bipy)2Cl](nbs) (1) (bipy = 2,2'-bipyridine, nbs = 4-nitrobenzenesulfonate) was obtained from the reaction of 4-nitrobenzenesulfonyl chloride and 2-amine-4-methylopyridine with CuCl2 in the presence of 2,2'-bipyridine and characterized by elemental analysis, IR spectra and X-ray single-crystal diffraction. The asymmetric unit of
Tsurng-Juhn Huang et al.
European journal of medicinal chemistry, 90, 428-435 (2014-12-03)
Hepatitis B virus (HBV) is a causative reagent that frequently causes progressive liver diseases, leading to the development of acute, chronic hepatitis, cirrhosis, and eventually hepatocellular carcinoma (HCC). Despite several antiviral drugs including interferon-α and nucleotide derivatives are approved for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service