Skip to Content
Merck
All Photos(1)

Key Documents

166634

Sigma-Aldrich

4,4′-Dibromobenzophenone

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$157.00

$157.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$157.00

About This Item

Linear Formula:
(BrC6H4)2CO
CAS Number:
Molecular Weight:
340.01
Beilstein:
2049958
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$157.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

powder

mp

171-174 °C (lit.)

SMILES string

Brc1ccc(cc1)C(=O)c2ccc(Br)cc2

InChI

1S/C13H8Br2O/c14-11-5-1-9(2-6-11)13(16)10-3-7-12(15)8-4-10/h1-8H

InChI key

LFABNOYDEODDFX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

4,4′-Dibromobenzophenone is the main degradation product of bromopropylate in honey[1].

Application

4,4′-Dibromobenzophenone was used in the synthesis of high molecular weight poly(p-phenylene) derivatives via palladium-catalyzed polycondensation reaction[2].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pd-catalyzed polycondensation of aromatic monomers with functional groups.
Rehahn M, et al.
Makromol. Chem., Rapid Commun., 11(11), 535-539 (1990)
G Formica
Journal - Association of Official Analytical Chemists, 67(5), 896-901 (1984-09-01)
An analytical procedure is described for determination of residues of bromopropylate (BP) and its main degradation product 4,4'-dibromobenzophenone (BBP) in honey. The method also allows the determination of 4,4'-dibromobenzilic acid (BBA), a potential intermediate to BBP. The method involves dissolution

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service