Skip to Content
Merck
All Photos(1)

Documents

378259

Sigma-Aldrich

4,4′-Diaminobenzophenone

97%

Synonym(s):

Bis(4-aminophenyl)methanone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(H2NC6H4)2CO
CAS Number:
Molecular Weight:
212.25
Beilstein:
2211861
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

solid

mp

243-247 °C (lit.)

SMILES string

Nc1ccc(cc1)C(=O)c2ccc(N)cc2

InChI

1S/C13H12N2O/c14-11-5-1-9(2-6-11)13(16)10-3-7-12(15)8-4-10/h1-8H,14-15H2

InChI key

ZLSMCQSGRWNEGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David R Worthen et al.
Bioorganic & medicinal chemistry letters, 19(17), 5012-5015 (2009-07-28)
Several diaminodiphenyl analogs were assessed in vivo for their capacity to inhibit seizure induction and propagation in rodents. Both 3,4'- and 4,4'-diaminodiphenyl compounds prevented seizures for as long as 4h after maximal electric shock induction. 4,4'-Diphenyl compounds bridged by a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service