Skip to Content
Merck
All Photos(1)

Documents

161160

Sigma-Aldrich

α-Bromocinnamaldehyde

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=C(Br)CHO
CAS Number:
Molecular Weight:
211.06
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

form

solid

mp

66-68 °C (lit.)

storage temp.

2-8°C

SMILES string

Br\C(C=O)=C/c1ccccc1

InChI

1S/C9H7BrO/c10-9(7-11)6-8-4-2-1-3-5-8/h1-7H/b9-6-

InChI key

WQRWNOKNRHCLHV-TWGQIWQCSA-N

General description

α-Bromocinnamaldehyde is commonly employed as an anti-mildew agent in commercial products.

Application

α-Bromocinnamaldehyde was used in the synthesis of 3,4-diaryl 1H-pyrazoles. It was also used in the preparation of spiro imidazolidine-oxazolidine intermediate via guanidinium ylide mediated aziridination.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Momma et al.
Eisei Shikenjo hokoku. Bulletin of National Institute of Hygienic Sciences, (107)(107), 29-36 (1989-01-01)
Bromocinnamic aldehyde (BCA), an antibacterial/antifungal agent, was tested for its chronic toxicity/carcinogenicity in female Slc:ddY mice. The animals received 0.25%, 1.0% and 4.0% of BCA dissolved in olive oil applied to the shaved back skin area twice a week for
Ya Zhang et al.
International immunopharmacology, 14(1), 107-113 (2012-06-20)
Early experiments showed cinnamaldehyde had obvious therapeutic effect on viral myocarditis, but cinnamaldehyde was unstable in vivo. To overcome this limitation, we used cinnamaldehyde as a lead compound to synthesize α-bromo-4-chlorocinnamaldehyde (BCC). In the present study, we compared the therapeutic
Wannaporn Disadee et al.
The Journal of organic chemistry, 71(17), 6600-6603 (2006-08-12)
We successfully isolated a spiro imidazolidine-oxazolidine intermediate in the reaction of guanidinium ylide mediated aziridination using alpha-bromocinnamaldehyde. X-ray crystallographic analysis unambiguously revealed that the stereogenic centers of the spiro intermediate were in a trans configuration. The role of the spiro
S Kojima et al.
Eisei Shikenjo hokoku. Bulletin of National Institute of Hygienic Sciences, (107)(107), 21-25 (1989-01-01)
The amount of alpha-bromocinnamaldehyde (BCA), an anti-mildew agent, in some commercial products, was examined by high performance liquid chromatography (HPLC) using the following conditions: column, Nucleosil 50-5 (Nagel, 250 mm x 4.6 mm i.d.); mobile phase, hexane-chloroform (12:5); flow rate
Yésica P Zaio et al.
Journal of the science of food and agriculture, 98(15), 5822-5831 (2018-05-15)
The insecticidal and repellent effects on adult Sitophilus zeamais of 12 cinnamaldehyde-related compounds was evaluated by contact toxicity bioassays and a two-choice olfactometer. To determine non-toxicity in mammals, body weight, serum biochemical profiles, liver weight, physiological parameters, sperm motility, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service