Skip to Content
Merck
All Photos(1)

Key Documents

15258

Sigma-Aldrich

Bis(triphenylphosphine)ruthenium(II) dicarbonyl chloride

≥98.0% (CH)

Synonym(s):

Dichlorodicarbonylbis(triphenylphosphine)ruthenium(II), Ruthenium(II)bis(triphenylphosphine)dichlorocarbonyl

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$299.00

$299.00


0 left in stock

Request a Bulk Order

Select a Size

Change View
1 G
$299.00

About This Item

Linear Formula:
[(C6H5)3P]2Ru(CO)2Cl2
CAS Number:
Molecular Weight:
752.57
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$299.00


0 left in stock

Request a Bulk Order

Assay

≥98.0% (CH)

form

powder

reaction suitability

core: ruthenium
reagent type: catalyst

SMILES string

Cl[Ru](Cl)(C#[O])(C#[O])([PH](c1ccccc1)(c2ccccc2)c3ccccc3)[PH](c4ccccc4)(c5ccccc5)c6ccccc6

InChI

1S/2C18H15P.2CO.2ClH.Ru/c2*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;2*1-2;;;/h2*1-15H;;;2*1H;/q;;;;;;+2/p-2

InChI key

PXURRFCLQDNZOY-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tao Jia et al.
Small (Weinheim an der Bergstrasse, Germany), 16(46), e2001343-e2001343 (2020-10-28)
Multimodal synergistic therapy based on photodynamic therapy (PDT), photothermal therapy (PTT), and chemodynamic therapy (CDT) has attracted increasing attention in cancer therapy. However, the scant therapeutic efficiency is always a barrier for further application. Herein, a smart tumor microenvironment (TME)

Questions

  1. where is the 1H ,13C and 31P NMR

    1 answer
    1. Unfortunately, NMR testing is not performed on this product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service