Skip to Content
Merck
All Photos(1)

Key Documents

225665

Sigma-Aldrich

Tris(triphenylphosphine)rhodium(I) carbonyl hydride

97%

Synonym(s):

RhH(CO)(PPh3)3, Rhodium monocarbonyl hydrogen tris(triphenylphosphine), Tris(triphenylphosphine)carbonylrhodium hydride, trans-Carbonyl(hydrido)tris(triphenylphosphine)rhodium, Carbonyltris(triphenylphosphine)rhodium(I) hydride, Hydridocarbonyl-tris(triphenylphosphine)-rhodium(I), Rhodium(I) tris(triphenylphosphine) carbonyl hydride

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$265.00
5 G
$1,600.00

$265.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$265.00
5 G
$1,600.00

About This Item

Linear Formula:
[(C6H5)3P]3Rh(CO)H
CAS Number:
Molecular Weight:
918.78
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$265.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

reaction suitability

core: rhodium
reagent type: catalyst

mp

150 °C (dec.) (lit.)

SMILES string

[RhH].[C-]#[O+].c1ccc(cc1)P(c2ccccc2)c3ccccc3.c4ccc(cc4)P(c5ccccc5)c6ccccc6.c7ccc(cc7)P(c8ccccc8)c9ccccc9

InChI

1S/3C18H15P.CO.Rh.H/c3*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;1-2;;/h3*1-15H;;;

InChI key

AACIZACVKFEETJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Catalyzes the ring-opening isomeriztion of methylenecyclopropanes to 1,3-dienes.[1]

Features and Benefits

Homogeneous catalyst.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J. Mol. Catal. A: Chem., 241, 65-65 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service